NucleoMag SEP
NucleoMag SEP
Actual product may vary
NucleoMag SEP
NucleoMag SEP
Fisherbrand
NucleoMag SEP
Magnetic separator, for use with 96‐well plates
Have Questions?
Catalog NumberQuantity
FKS119925821 Ea.
Catalog number FKS11992582
Price (SGD)
-
Quantity:
1 Ea.
NucleoMag™ kits are based on Macherey-Nagel's magnetic-bead technology and allow for rapid simultaneous purification of high-quality nucleic acids from whole blood. The magnetic bead technology provides a closed system, greatly reducing the risk for cross-contamination. The kits and Separator can also be used on common liquid handling instruments, allowing complete automation.
Using the NucleoMag™ method, genomic DNA is selectively bound to the NucleoMag™ Beads. During washing and elution, the beads are freely dispersed in solution. To separate the beads from the solution, the separation plate is transferred onto the NucleoMag™ SEP. The beads are attracted to the magnet. Meanwhile, the wash buffer can be removed from the well or the elution buffer containing the purified DNA can be recovered, respectively.
  • Magnetic bead technology
  • Yield: 1μg to 4μg
  • Sample material: 100μL whole blood
  • Elution volume: 25μL to 100μL
  • Binding capacity: 0.4μL/1μL beads
  • Preparation time: 120 min./96 preps
  • Fragment size: 300bp to 30kb
  • Format: superparamagnetic beads (size of beads: 1μm to 3μm)
Specifications
Isolation TechnologyMagnetic Bead
For Use With (Equipment)NucleoMag™ 96 Kits
For Use With (Application)Rapid manual purification of nucleic acids
Product TypeNucleoMag™ SEP Magnetic Separator
Quantity1 Ea.
Unit SizeEA
Contents & Storage
1 Ea., 15°C to 25°C