Printed with pin sharp blue graduations for enhanced visibility. Azlon™ Azlon™ Polypropylene Printed Jugs provide a reliable solution for measuring and dispensing liquids in a laboratory.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS11990985
3000 mL
FKS10667825
250 mL
FKS10074084
500 mL
FKS11363434
1000 mL
FKS10718205
2000 mL
FKS10617065
5000 mL
6 Options
Catalog number FKS11990985
Price (SGD)
56.00
EA
Add to cart
Capacity (Metric):
3000 mL
Price (SGD)
56.00
EA
Add to cart
Printed with pin sharp blue graduations for enhanced visibility. Azlon™ Azlon™ Polypropylene Printed Jugs provide a reliable solution for measuring and dispensing liquids in a laboratory.
Tapers: to form a large pouring spout
Constructed: from high clarity polypropylene for strength and visibility
Graduations: printed enhanced pin sharp blue
Autoclavable: to 121°C
Ergonomically designed: handle with thumb grip for easy carrying and pouring
Suitable: for food contact applications per EU and US regulations
Specifications
ColorClear
Diameter (Metric) Outer172 mm
Diameter (Metric)Top172 mm
Capacity (Metric)3000 mL
MaterialPolypropylene
For Use With (Application)Laboratories for Handling Chemicals
Height (Metric)215 mm
Quantity1/Pk.
Temperature (Metric) Max. Operating-20°C to +135°C