Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Height (Metric) | Length (Metric) Stem |
---|---|---|---|
FKS11987904 | 225 mL | 17.7 cm | 9.9cm |
FKS11977914 | 50 mL | 10.9 cm | 6.5cm |
FIS10-500-13 | 37 mL | 10.2 cm | 6.1cm |
FKS11967914 | 37 mL | 10.2 cm | 6.1cm |
FKS11947914 | 12 mL | 7.5 cm | 5.2cm |