Check Order Status
Soda Lime Glass Rimless Test Tube
Actual product may vary
Fisherbrand
Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11982188
75 mm
10 mm
Catalog number
FKS11982188
Price (SGD)
-
Length (Metric):
75 mm
Diameter (Metric):
10 mm
Soda Lime Glass
Rimless
Disposable
Specifications
Certifications/Compliance
ISO 4142
Length (Metric)
75 mm
Material
Soda Lime Glass
Quantity
100 Pack
Rimmed
No
Diameter (Metric)
10 mm