Bulb Pipettes, One Mark, Class B, BS 1583
Actual product may vary
Fisherbrand
Bulb Pipettes, One Mark, Class B, BS 1583
Pipette with precision, Fisherbrand one mark Class B pipettes are made of soda lime glass. Colour coded with blue markings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color
FKS1198216850 mLRed
FKS1196216810 mLRed
FKS1197216825 mLBlue
FKS119521685 mLWhite
Catalog number FKS11982168
Price (SGD)
-
Volume (Metric):
50 mL
Color:
Red
  • Color coded to BS 3996
  • Class B
  • Soda Lime Glass
  • Graduated
  • One Mark
  • Type 1
Specifications
CalibrationTo Deliver
Certifications/ComplianceISO 648, DIN 12690, Class B
ClassB
MaterialSoda Lime Glass
GraduatedYes
Quantity2 Pack
ColorRed
Volume (Metric)50 mL
TypeBulb Pipet
Unit SizePack