Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Socket Joint
FKS11980885250 mL130 mm24/29
FKS10243930150 mL110 mm24/29
Catalog number FKS11980885
Price (SGD)
543.00
Pack
Capacity (Metric):
250 mL
Height (Metric):
130 mm
Socket Joint:
24/29
Price (SGD)
543.00
Pack
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
  • Complies with ISO 1773 and ISO 4797
  • With highest quality Quickfit™ ground glass socket
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Flat base allows use with hotplates
Specifications
Certifications/ComplianceISO 1773, ISO 4797
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (Metric)130 mm
Neck StyleShort
Socket Joint24/29
Bottom ShapeFlat
Unit SizePack