Disposable Slide Staining Tray
Disposable Slide Staining Tray
Actual product may vary
Fisherbrand
Disposable Slide Staining Tray
Stain, rinse and dry slides on one working tray. Disposable Slide Staining Tray holds eight slides with deep wells. Rugged, economical trays provide a clean, fresh working service every time. Includes a dark lid, pour spouts and recessed handles for superior handling.
Have Questions?
Catalog NumberProduct Type
FKS11968004Slide Staining Tray
Catalog number FKS11968004
Price (SGD)
189.00
PK
Add to cart
Product Type:
Slide Staining Tray
Price (SGD)
189.00
PK
Add to cart
  • Disposable trays are an economical alternative to metal slide trays.
  • Reduce possible cross contamination and keep work areas neat.
  • Dark lid protects slides for light-sensitive applications.
  • Compact size and recessed handles for easy transport from work area to sink.
  • Two convenient pour spouts for quick disposal of unwanted liquid waste.
Specifications:
  • Dimensions: 305 x 127 x 39.5 mm
  • Well capacity: 38 mL
  • Color: Black
  • Materials: Polypropylene and polyethylene blend
Specifications
Product TypeSlide Staining Tray
Slide Capacity8
MaterialPolypropylene, Polyethylene
Media Capacity38 mL
ColorBlack
Includes4 Trays and 1 Dark Lid
Quantity4 Trays, 1 Lid
Dimensions (L x W x H)305 x 127 x 39.5 mm
Unit SizePK