CETI 4.5kg Convex Refractometer
CETI 4.5kg Convex Refractometer
Actual product may vary
Fisherbrand
CETI 4.5kg Convex Refractometer
Highly versatile refractometer that allows measurements of a wide range of substances. Medline Scientific™ CETI 4.5kg Convex Refractometer is capable of reading both reflected and transmitted light.
Have Questions?
Catalog number FKS11965518
Price (SGD)
9,857.00
EA
Add to cart
Price (SGD)
9,857.00
EA
Add to cart
  • Wide range of measurable substances
  • Light entry ports for both upper and lower prisms
  • Supplied with a calibration set
    • Glass test piece
    • Alpha-Bromonaphtalene
    • Screwdriver
  • Supplied with dispersion tables, instruction manual, dust cover
Specifications
TypeConvex Refractometer
Scale Range0 to 95 Brix
Refractive Index1.3000 to 1.7200
Resolution±0.1°C
Includes30x focusable telescope eyepiece, simultaneous scale reading, and border line setting
Quantity1 Each
Weight (Metric)4.5 kg
Dimensions (L x W x H)280 x 190 x 380 mm
Height (Metric)380 mm
Length (Metric)280 mm
Temperature Range (Metric)0°C to 200°C
Unit SizeEA