1x1 image pixel for data collection
NucleoSpin™ Soil, Mini kit for DNA from soil
NucleoSpin™ Soil, Mini kit for DNA from soil
Actual product may vary
NucleoSpin™ Soil, Mini kit for DNA from soil
NucleoSpin™ Soil, Mini kit for DNA from soil
NucleoSpin™ Soil, Mini kit for DNA from soil
Fisherbrand
NucleoSpin™ Soil, Mini kit for DNA from soil
Isolate DNA from soil. Macherey Nagel™ NucleoSpin™ enables soil Isolation of total DNA from diverse soil types
Have Questions?
Catalog NumberNo. of Reactions
FKS1196240250 Preps
Catalog number FKS11962402
Price (SGD)
-
No. of Reactions:
50 Preps
Features
  • Two alternative lysis buffers and a special additive (Enhancer SX) for optimal processing of various soil samples – high yield and high purity
  • NucleoSpin™ Bead Tubes with ceramic beads for most efficient cell disruption of Gram-positive and Gram-negative bacteria, archaea, yeast, fungi, and algae in soil, sludge,or sediment
  • NucleoSpin™ Inhibitor Removal Column to eliminate all PCR inhibitors, such as humic substances – DNA is ready-to-use for PCR without dilution
Applications
  • Total DNA from microorganisms in soil and sediment
  • Quantify and detect microorganisms from environmental samples
  • Typical downstream applications: PCR, real-time PCR, Southern blotting, microarray technology
Specifications
FormatMini Spin Column
Isolation TechnologySilica Membrane Technology
For Use With (Application)For Total DNA From Microorganisms in Soil and Sediment
No. of Reactions50 Preps
Sample Volume<500 mg
Test Time90min.
Yield2 to 10μg
Unit SizeEA
Share catalog number, name or link