Check Order Status
Soda Lime Glass Light Wall Rimmed Test Tube
Actual product may vary
Fisherbrand
Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
Length (Metric)
FKS11962188
16 mm
150 mm
FKS11952188
16 mm
125 mm
2 Options
Catalog number
FKS11962188
Price (SGD)
73.00
PK
Add to cart
Diameter (Metric):
16 mm
Length (Metric):
150 mm
Price (SGD)
73.00
PK
Add to cart
Withstands moderate warming
Light weight
Complies with ISO 4142
Specifications
Certifications/Compliance
ISO 4142
Length (Metric)
150 mm
Material
Soda Lime Glass
Quantity
100 Pack
Rimmed
Yes
Diameter (Metric)
16 mm