1x1 image pixel for data collection
Glass Funnel Filter with Sintered Glass Disc
Glass Funnel Filter with Sintered Glass Disc
Actual product may vary
Fisherbrand
Glass Funnel Filter with Sintered Glass Disc
Constructed of borosilicate glass and available in a variety of capacities. Fisherbrand™ Glass Funnel Filters with Sintered Glass Disc filter medium precipitates ranging from 16 - 40μm and 10 to 14μm pore sizes respectively.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)PorosityDiameter (Metric) Disc
FKS11962148500 mL395mm
FKS11952148125 mL465mm
Catalog number FKS11962148
Price (SGD)
435.00
PK
Add to cart
Capacity (Metric):
500 mL
Porosity:
3
Diameter (Metric) Disc:
95mm
Price (SGD)
435.00
PK
Add to cart
Constructed of borosilicate glass and available in a variety of capacities. Fisherbrand™ Glass Funnel Filter with Sintered Glass Disc filters medium precipitates ranging from 16 - 40μm and 10 to 14μm pore sizes respectively.
  • Funnels constructed of borosilicate glass.
  • Funnel filters constructed of sintered glass.
  • Filters medium precipitates with grade 3 and grade 4 porosity.
Specifications
Capacity (Metric)500 mL
MaterialBorosilicate Glass
Diameter (Metric) Disc95mm
IncludesSintered glass disc
Porosity3
Product TypeFilter Funnel
Unit SizePK
Share catalog number, name or link