Cylindrical Jars with Ribbed Cap
Actual product may vary
Cylindrical Jars with Ribbed Cap
Manufactured from HDPE for optimum strength in a lightweight container. Kartell™ Cylindrical Jars with Ribbed Cap offer an economical jar with low environmental impact for powders, pastes and solids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11959848500 mL
FKS12543197250 mL
FKS123685501000 mL
Catalog number FKS11959848
Price (SGD)
327.00
Pack
Capacity (Metric):
500 mL
Price (SGD)
327.00
Pack
Manufactured from HDPE for optimum strength in a lightweight container. Kartell™ Cylindrical Jars with Ribbed Cap offer an economical jar with low environmental impact for powders, pastes and solids.
  • Exceptionally wide mouth bottles with insert and cap
  • Ribbed cap for improved grip
  • Leakproof
  • Suitable for foodstuff per Framework Regulation (EC) 1935/2004 and Commission Regulation (EU) 10/2011
Specifications
Capacity (Metric)500 mL
Diameter (Metric) Inner Neck70 mm
For Use With (Application)Foodstuff, Powders, Solid Specimen, Creamy or Semisolid Substances, Crystals and Hygroscopic Substances
IncludesInsert and Ribbed Cap
MaterialHDPE
No. per Pack50 Pk.
Certifications/ComplianceCE and IVD, per EC Regulation 1935/2004, EC Regulation 10/2011
Closure IncludedYes
MouthWide
ShapeRound
Height (Metric)107 mm
Product TypeJar
Diameter (Metric)86 mm
Closure TypeRibbed
Unit SizePack