Cargo™ Pipet Rack
Cargo™ Pipet Rack
Actual product may vary
Fisherbrand
Cargo™ Pipet Rack
Configure the bins to match available benchtop surface. Cargo™ Pipet Rack can be connected in multiple ways or used individually to meet workspace and storage needs. Sturdy ABS plastic bins with angled shelves and protective lids keep pipets safely within arm’s reach.
Have Questions?
Catalog NumberColor
FKS11958004Blue
Catalog number FKS11958004
Price (SGD)
370.00
PK
Add to cart
Color:
Blue
Price (SGD)
370.00
PK
Add to cart
  • Ideal for storing and organizing 1.0 mL to 50 mL serological pipets
  • Use as individual holders or connect multiple units for larger storage needs
  • Click together horizontally or vertically to build the desired configuration
  • Protective lids enclose the units to prevent dust build-up and provide additional shelf space
  • Magnets on both sides of the unit connect securely and safely to metal surfaces
Specifications
IncludesLid
Product TypePipette Rack
AutoclavableNot Autoclavable
ColorBlue
HoldsPipettes
MaterialABS
Positions1
ShapeRectangular
ShelvesAngled
StackableYes
SterilityNon-sterile
For Use With (Application)Ideal for Storing and Organizing Pipets
For Use With (Equipment)Serological Pipets
Quantity3 Pk.
Dimensions (L x W x H)301 x 100 x 127.5 mm
Length (Metric)301 mm
Length (English)11.9 in.
Width (Metric)100 mm
Width (English)3.9 in.
Height (Metric)127.5 mm
Height (English)5 in.
Unit SizePK