Check Order Status
Maxi Pipet Tips
Actual product may vary
Fisher Catalogue
Maxi Pipet Tips
For use with large volume pipets
Have Questions?
Change view
Catalog Number
Length (Metric)
FKS11957754
152 mm
FIS02-681-412
161 mm
FKS11947754
161 mm
FIS02-707-466
152 mm
4 Options
Catalog number
FKS11957754
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
152 mm
For use with large volume pipets
Designed for use when transferring large sample volume
Advanced molding technology ensure tips are straight
Alternative option to serological pipets
Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications
Volume (Metric)
1 to 10 mL
Filtered
Non-filtered
Sterility
Non-sterile
Color
Clear
Autoclavable
Autoclavable
Length (Metric)
152 mm
Format
Bulk
Tip Style
Macro
For Use With (Equipment)
Fisherbrand™ II, Thermo Scientific™
Product Line
Maxi
Material
Polypropylene