Legato 100 Infuse Only Syringe Pump
Legato 100 Infuse Only Syringe Pump
Actual product may vary
Fisherbrand
Legato 100 Infuse Only Syringe Pump
Versatile pump offers unparalleled ease of use through simple touch-screen interface
Have Questions?
Catalog number FKS11955596
Price (SGD)
13,024.00
EA
Add to cart
Price (SGD)
13,024.00
EA
Add to cart
Versatile pump offers unparalleled ease of use through simple touch-screen interface
Infuse only single syringe pump. Accommodates syringes from 0.5μL to 60mL. User definable flowrates with selectable target volume or time values to control the total infusion volume.
  • Holds one syringe from 0.5μL to 60mL
  • High resolution color touch screen
  • Touch pad lock feature
  • LED light on front panel
  • Full metal chassis
  • Built-in syringe table
  • Up to 30 lb. linear force
  • Advanced microstepping techniques
  • Built-in RS485 interface
  • USB port
  • I/O and TTL interface
  • Continuous mode of operation
  • Spill dam
Specifications
  • Accuracy: ±0.5 %
  • Reproducibility: ±0.05%
  • Syringes (min/max): 0.5μL to 60mL
  • Flow Rate, Minimum (0.5μL syringe): 1.28 pL/min.
  • Flow Rate, Maximum (10mL syringe): 25.99 mL/min.
  • Flow Rate, Maximum (60mL syringe): 88.28 mL/min.
  • Display: 4.3 WQVGA TFT Color Display with touchpad
  • Non-Volatile Memory: Stores all settings
  • Operating Temperature: 4°C to 40°C (40°F to 104°F)
  • Storage Temperature: -10°C to 70°C (14°F to 158°F)
  • Humidity: 20% to 80% RH noncondensing
  • Mode of Operation: Continuous
  • Power: 100-240 VAC: 50/60 Hz, 50W, 0.5A fuse
  • Dimensions: 9 x 7.5 x 5 in. (22.6 x 19.05 x 15cm)
  • Weight: 5.9 lb. (2.66kg)
Specifications
Voltage100/240 V
Flow Rate1.28 pl/min. to 88.28 mL/min.
Capacity (Metric)0.5 μL to 60 mL
Certifications/ComplianceCE, UL, CSA, CB Scheme, EU RoHS
ConnectorsRS485 - IEEE-1394 6 pos, USB - Type B, I/O and TTL - 15 pin D-sub connector
ControllerMicroprocessor with 0.062 microstepping
Linear Force (Metric)13.6 kg
Linear Force (English)30 lb.
MaterialMetal
Reproducibility±0.05%
Temperature (Metric) Storage-10°C to +70°C
TypeInfusion Syringe Pump
Weight (English)5.9 lb.
Weight (Metric)2.66 kg
Electrical Requirements100/240 V 50/60 Hz
Display TypeTFT
Flow Rate Accuracy±0.5%
Number of Channels1
Frequency50/60 Hz
Overall Height (English)5 in.
Overall Height (Metric)15 cm
Overall Length (English)9 in.
Overall Length (Metric)7.5 cm
Overall Width (English)7.5 in.
Temperature (Metric) Max40°C
Temperature (Metric) Min.4°C
Unit SizeEA