SureOne™ Micropoint Pipette Tips, Universal Fit, Non-Filtered
Actual product may vary
Fisherbrand
SureOne™ Micropoint Pipette Tips, Universal Fit, Non-Filtered
New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityTip StyleFormat
FKS119534160.1 to 10 μLNon-sterileMicro PointHinged Rack
FIS02-707-400100 to 1250 μLNon-sterileMicro PointBulk
FKS11548442100 to 1250 μLNon-sterileMicro PointBulk
FIS02-707-407100 to 1250 μLSterileMicro PointHinged Rack
FIS02-707-4105 to 300 μLSterileBeveledHinged Rack
FIS02-707-4475 to 300 μLNon-sterileMicro PointBulk
FKS115384425 to 300 μLNon-sterileBeveledRefill
FIS02-707-4360.1 to 10 μLNon-sterileMicro PointHinged Rack
FIS02-707-4400.1 to 10 μLNon-sterileExtended LengthHinged Rack
FIS02-707-4410.1 to 10 μLSterileExtended LengthHinged Rack
FIS02-707-4540.1 to 10 μLNon-sterileMicro PointBulk
FIS02-707-4370.1 to 10 μLSterileMicro PointHinged Rack
Catalog number FKS11953416
Price (SGD)
129.00
Pack
Volume (Metric):
0.1 to 10 μL
Sterility:
Non-sterile
Tip Style:
Micro Point
Format:
Hinged Rack
Price (SGD)
129.00
Pack
Reload system provides a complete line of micropoint universal-fit pipet tips.
  • All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors and endotoxin and DNA All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
SureOne Rack Features
  • Flush Fit Inserts sit flush with rack, allowing easy multichannel pipetting
  • Recycle Logo molded into all rack components
  • Notched clasp props lid open for autoclave venting
  • Sure Stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe-Lock latch allows for one-handed open and closing ability and won't pop open when dropped
  • Sure-snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Reversible flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
SureOne Insert Features
  • Inserts are easy to place onto and remove from rack; clip onto rack for a secure fit
  • Lot traceable—all inserts contain the Fisherbrand part number, tip volume, lot number
  • Indented insert holes inhibit tip movement, stabilizing them for multichannel work
  • Contact Clip prevents unnecessary contact with the tips while clipping in inserts
  • Multichannel design; all inserts are 96-well format Robust non-pliable design provides secure seals on every tip
SureOne Reload Packaging Features
  • Product information displayed on box
  • Dust Flap prevents debris from contaminating the product
  • Finger Knockouts allow easy access to slide out product
  • Canyon Cut Package allows for easy tip insert removal
  • Reload packaging design saves 58% bench space vs. racked packaging; reduces plastic waste by 52%
As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.
Specifications
Volume (Metric)0.1 to 10 μL
FilteredNon-filtered
SterilityNon-sterile
ColorClear
Length (English)1.25 in.
Length (Metric)31 mm
Clean ClaimsDNase, RNase, and DNA Free
FormatHinged Rack
Tip StyleMicro Point
For Use With (Equipment)Universal Fit
GraduatedYes
Product LineSureOne
MaterialPolypropylene
Graduations2μL
Unit SizePack