▸
Baths
▸
Bath Accessories
▸
Aqua Stabil™ Water Bath Protective Media
Actual product may vary
Fisherbrand
Aqua Stabil™ Water Bath Protective Media
Julabo™ Aqua Stabil™ Water Bath Protective Media stops growth of algae, bacteria and other microorganisms with its bactericidal effectiveness.
Have Questions?
Catalog Number
Type
FKS11950218
Aqua Stabil Protective Media
Catalog number
FKS11950218
Price (SGD)
1,490.00
Pack
Add to cart
Type:
Aqua Stabil Protective Media
Price (SGD)
1,490.00
Pack
Add to cart
Protects bath fluid
Provides long period of use for fluid
Specifications
Description
6 Bottles
For Use With (Equipment)
ED, ED-13, ED-13A, ED-13M, ED-17 Open heating bath circulator, Water bath
Type
Aqua Stabil Protective Media