Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Actual product may vary
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Fisher Catalogue
Polypropylene Copolymer (PPCO) Wide-Mouth Bottles
Higher temperature resistance than LDPE or HDPE
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS11947934250 mL45 mm
FIS02-896B60 mL28-415
FIS02-896C125 mL38 mm
FKS11937934125 mL38 mm
FIS02-896D250 mL45 mm
FIS02-896E500 mL55 mm
FIS02-896F1000 mL65 mm
Catalog number FKS11947934
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Size:
45 mm
Higher temperature resistance than LDPE or HDPE
  • Handle up to 120°C (248°F)† for short periods and 100°C (212°F) continuously
  • Brittleness temperature of 0°C (32°F)
  • Rigid walls
  • Excellent chemical resistance
  • Can be sterilized by all normal methods, except dry heat and radiation
Completely disengage threads or remove cap before autoclaving.
Specifications
Capacity8.45 oz., 250mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
MaterialPPCO
SterilityNon-sterile
Neck Finish45mm
Certifications/ComplianceISO 13485:2003; QMI-SAI File No.1606319 and 1606321
Closure IncludedYes
AutoclavableAutoclavable
MouthWide
Product TypeWide Mouth Bottle
Closure LinerUnlined
Closure MaterialPPCO
Closure Size45 mm
Closure TypeScrew Cap
Unit SizePK