Check Order Status
Borosilicate Glass Test Tube
Actual product may vary
Fisherbrand
Borosilicate Glass Test Tube
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11942198
75 mm
12 mm
Catalog number
FKS11942198
Price (SGD)
-
Length (Metric):
75 mm
Diameter (Metric):
12 mm
Borosilicate glass test tubes, grade 3.3
All ISO 4142
Specifications
Certifications/Compliance
ISO 4142
Length (Metric)
75 mm
Material
Borosilicate Glass
Quantity
100 Pack
Diameter (Metric)
12 mm