Metal Stop-it Hose Clamp
Metal Stop-it Hose Clamp
Actual product may vary
Fisherbrand
Metal Stop-it Hose Clamp
Buerkle™ Metal Stop-it Hose Clamp can be used wherever the flow in hoses with liquids or gases needs to be regulated or stopped.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS1194160130 mm O.D. Flexible Hoses
Catalog number FKS11941601
Price (SGD)
44.00
EA
Add to cart
For Use With (Equipment):
30 mm O.D. Flexible Hoses
Price (SGD)
44.00
EA
Add to cart
  • Galvanised steel and aluminum
  • For flexible hoses with outer Ø up to 10mm, 15mm, 20mm and 30mm
  • Ensure that the stop-it metal is extremly heat resistant and sturdy
  • With the hose clip stop-it's knurled screw, a stepless, delicate flow regulation or complete stop of the flow are possible, even for existing hose connections
  • The hose is compressed over a wide area against the sturdy steel hook in order to prevent it from being damaged
Specifications
TypeHose Clamp
MaterialGalvanized Steel, Aluminum
For Use With (Equipment)30 mm O.D. Flexible Hoses
DescriptionExtremely heat resistant and sturdy
Unit SizeEA