Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm
Actual product may vary
Pyrex
Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm
Manufactured from Borosilicate Glass and complies with ISO 6556. Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm features heavy walls to provide increased mechanical strength while 'studded' base minimises surface abrasion. These Büchner type flasks include an integral glass side-arm to accept 9mm bore flexible tubing necessary for vacuum filtration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Inner NeckDiameter (Metric) Outer
FKS11941548250 mL155 mm34 mm85 mm
FKS120898972000 mL260 mm60 mm166 mm
FKS125599653000 mL335 mm68 mm187 mm
FKS113878345 L385 mm68 mm220 mm
Catalog number FKS11941548
Price (SGD)
1,463.00
Pack
Capacity (Metric):
250 mL
Height (Metric):
155 mm
Diameter (Metric) Inner Neck:
34 mm
Diameter (Metric) Outer:
85 mm
Price (SGD)
1,463.00
Pack
Manufactured from Borosilicate Glass and complies with ISO 6556. Pyrex™ Borosilicate Glass Vacuum Filter Flask with Side Arm features heavy walls to provide increased mechanical strength while 'studded' base minimises surface abrasion. These Büchner type flasks include an integral glass side-arm to accept 9mm bore flexible tubing necessary for vacuum filtration.
  • Complies with ISO 6556
  • Büchner type flasks for vacuum filtration
  • Heavy walls provide increased mechanical strength
  • Studded base to minimise surface abrasion
  • With integral glass sidearm to accept 9mm bore flexible tubing
Specifications
Certifications/ComplianceISO 6556
DescriptionStyle: Erlenmeyer
Diameter (Metric) Inner Neck34 mm
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Height (Metric)155 mm
Diameter (Metric) Outer85 mm
Closure Size33
Unit SizePack