Polypropylene Low Form Beakers with Molded Graduations
Polypropylene Low Form Beakers with Molded Graduations
Actual product may vary
Fisher Catalogue
Polypropylene Low Form Beakers with Molded Graduations
Manufactured from highly translucent polypropylene for durability, clarity and good chemical resistance. Autoclavable Kartell™ Polypropylene Low Form Beakers with Molded Graduations provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS119398582000 mL
FKS11919858500 mL
FKS119298581000 mL
Catalog number FKS11939858
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Manufactured from Polypropylene for durability and good chemical resistance. Autoclavable Kartell™ Polypropylene Class B Low Form Beakers provide an economical and safe solution for stirring, mixing and dispensing liquids in the laboratory.
  • Permanent molded graduations provide reproducibility and clarity
  • Autoclavable at 121°C for 10 minutes
  • Withstands 100°C of continuous work
  • Class B, Low form
  • Complies with ISO 7056-1981 (E) and BS 5404 Part 1
  • Suitable for foodstuff
Specifications
Diameter (Metric) Outer132 mm
Capacity (Metric)2000 mL
MaterialPolypropylene
For Use With (Application)Suitable for Foodstuff
Height (Metric)183.5 mm
Increments50 mL
Temperature (Metric) Max. Operating100°C for continuous work
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceISO 7056-1981(E), BS 5404 Part 1
Unit SizePK