Store, dispense and cut sealing film. Fisherbrand™ Acrylic Parafilm™ Dispenser features a serrated cutting blade to easily slice film. Sturdy dispenser holds 50mm or 100mm rolls without cluttering your workspace. Clear sides make it easy to monitor inventory level.
Have Questions?
Catalog Number
Color
FKS11937994
Clear
Catalog number FKS11937994
Price (SGD)
457.00
EA
Add to cart
Color:
Clear
Price (SGD)
457.00
EA
Add to cart
Heavy-duty construction for increased durability
Ideal for Parafilm™, tape, labels and Tough-Tags™
Strong, crystal-clear acrylic dispenser with a serrated cutting blade
Holds two 50mm rolls side-by-side or one 100mm roll
Specifications
Product TypeParafilm Dispenser
ColorClear
Length (Metric)168 mm
Width (Metric)119 mm
Height (Metric)175 mm
MaterialAcrylic
Capacity2 x 50 mm rolls or 1 x 100 mm roll
For Use With (Equipment)Parafilm™ Sealing Film rolls