Azlon™ Polypropylene Test Tube Rack
Azlon™ Polypropylene Test Tube Rack
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Test Tube Rack
Economical and sturdy test tube racks supplied in self assembly flat packs
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositionsDiameter (Metric) OpeningDimensions (L x W x H)
FKS11933849Blue2131 mm265 x 126 x 88 mm
FKS11397794White3226 mm265 x 126 x 88 mm
Catalog number FKS11933849
Price (SGD)
448.00
PK
Add to cart
Color:
Blue
Positions:
21
Diameter (Metric) Opening:
31 mm
Dimensions (L x W x H):
265 x 126 x 88 mm
Price (SGD)
448.00
PK
Add to cart
Economical and sturdy test tube racks supplied in self assembly flat packs
  • Autoclavable at 121°C
  • Sinks in water baths
  • Alpha numeric place coding
Specifications
Product TypeTest Tube Rack
AutoclavableAutoclavable
ColorBlue
HoldsTubes (30 to 31 mm)
MaterialPolypropylene
Positions21
Dimensions (L x W x H)265 x 126 x 88 mm
Diameter (Metric) Opening31 mm
Diameter (Metric) Inner31 mm
Unit SizePK