BMP™51/BMP™53 Label Maker Cartridges: CleanLift™ Indoor/Outdoor Vinyl
BMP™51/BMP™53 Label Maker Cartridges: CleanLift™ Indoor/Outdoor Vinyl
Actual product may vary
Fisherbrand
BMP™51/BMP™53 Label Maker Cartridges: CleanLift™ Indoor/Outdoor Vinyl
BMP™51/BMP™53 Cartridges: CleanLift™ Vinyl labels are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHeight (Metric)Width (Metric)
FKS11932872Black on White762 cm38.10 mm
FIS19-155-172Black on White762 cm38.10 mm
FIS19-155-170Black on White762 cm12.70 mm
Catalog number FKS11932872
Price (SGD)
277.00
BOX
Add to cart
Color:
Black on White
Height (Metric):
762 cm
Width (Metric):
38.10 mm
Price (SGD)
277.00
BOX
Add to cart
BMP™51/BMP™53 Cartridges: CleanLift™ Vinyl labels are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
  • All-in-one cartridge includes ribbon and label supply
Specifications
ColorBlack on White
For Use With (Equipment)BBP31 Sign and Label Printer, GlobalMark Industrial Label Maker, MiniMark Industrial Label Printer, PowerMark Sign and Label Printer, Labelizer Plus Sign and Label Maker
MaterialVinyl
Length (English)25 ft.
Width (English)1.5 in.
Width (Metric)38.10 mm
Height (Metric)762 cm
Thickness (English)0.004 in.
Adhesive TypePermanent
SterilityNon-sterile
FinishGloss
Temperature Range (English)-40 to 180°F
Temperature Range (Metric)-40°C to +82°C
Certifications/ComplianceASTM D 523, PSTC-133, RoHS
For Use With (Application)Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
Quantity1 Ea.
Unit SizeBOX