Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Actual product may vary
Fisherbrand
Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.45 μm Pore Size
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Have Questions?
Catalog NumberSurface TypeSterilityIncludesPackaging
FKS11929207GridSterileInterleaving paperIndividually Packed
Catalog number FKS11929207
Price (SGD)
896.00
PK
Add to cart
Surface Type:
Grid
Sterility:
Sterile
Includes:
Interleaving paper
Packaging:
Individually Packed
Price (SGD)
896.00
PK
Add to cart
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
  • Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
  • Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
  • Special nontoxic ink used is completely free from bacterial growth inhibitors
  • Manufactured under strictly controlled cleanroom conditions
Specifications
AutoclavableAutoclavable
ColorWhite
Material (Membrane)MCE
Pore Size0.45 μm
SterilitySterile
For Use With (Application)Microbiological work, sample preparation, microbiological studies, filtration of aqueous solutions
Grid ColorBlack
IncludesInterleaving paper
PackagingIndividually Packed
Protein Binding CapacityMedium
Quantity200Pack
Size (Metric) Grid3.1mm
Surface TypeGrid
Temperature (English) Operating266°F
Temperature (Metric) Operating130°C
Diameter (English)1.85 in.
Diameter (Metric)47 mm
Unit SizePK