Professional Plus Multiparameter Meter
Professional Plus Multiparameter Meter
Actual product may vary
Professional Plus Multiparameter Meter
Professional Plus Multiparameter Meter
Fisherbrand
Professional Plus Multiparameter Meter
Versatile handheld instrument for pH, DO, conductivity and ISE measurements
Have Questions?
Catalog NumberMeasurement Modes
FKS11918089pH, Dissolved Oxygen, Conductivity, ISE
Catalog number FKS11918089
Price (SGD)
-
Measurement Modes:
pH, Dissolved Oxygen, Conductivity, ISE
Compact, handheld instrument provides true versatility in a multiparameter platform. Simply choose the cable length and probes to suit your application; then determine what you want the instrument to display.
  • Two mV inputs for combinations of Ammonium, Chloride, Nitrate, pH and ORP.
  • Choice of lab or field grade pH and ORP probes
  • IP67 waterproof rating
  • Choice of 1, 4, 10, 20, and 30m (3, 13, 32, 65 and 98 ft.) detachable cables (available separately)
  • Automatic buffer recognition
  • Automatic temperature, salinity, and barometric compensation for dissolved oxygen measurements
  • Continuous battery level display
  • Backlit display and keypad
  • User-upgradeable software via USB communications
  • Graphic display with detailed messages and instructions
  • 2000 data-set memory with additional GLP event logging
  • Interval or single event logging capability
  • Data Manager desktop software to manage all your data
Specifications
Measurement ModespH, Dissolved Oxygen, Conductivity, ISE
IncludesMeter, two alkaline C-cells, instructions, communications saddle, USB cable and YSI Data Manager software for Windows.
Unit SizePK