Plastilab™ Unequal Straight Tapered Tubing Connectors
Plastilab™ Unequal Straight Tapered Tubing Connectors
Actual product may vary
Fisherbrand
Plastilab™ Unequal Straight Tapered Tubing Connectors
Designed with different inlet and outlet diameters.
Have Questions?
Catalog NumberPort Size
FKS11914289Bore Size: 1.6 to 4.6mm
Catalog number FKS11914289
Price (SGD)
53.00
PK
Add to cart
Port Size:
Bore Size: 1.6 to 4.6mm
Price (SGD)
53.00
PK
Add to cart
Designed with different inlet and outlet diameters.
  • For use with flexible tubing
  • Autoclavable
Specifications
Diameter (Metric)4-8 mm to 10-12 mm
AutoclavableAutoclavable
MaterialPolypropylene
Diameter (English) Outer0.15 to 0.31 in., 0.39 to 0.47 in.
Diameter (Metric) Outer4 to 8 mm, 10 to 12 mm
For Use WithFor flexible tube
Packaging Quantity10/Pk.
Port SizeBore Size: 1.6 to 4.6mm
ShapeStraight
FlexibilityFlexible
Product TypeUnequal Straight Tapered Tubing Connector
Unit SizePK