Sample Storage Assembled Screw Vial Kits
Sample Storage Assembled Screw Vial Kits
Actual product may vary
Sample Storage Assembled Screw Vial Kits
Sample Storage Assembled Screw Vial Kits
Fisherbrand
Sample Storage Assembled Screw Vial Kits
Store your samples with confidence and convenience with these assembled screw vial kits.
Have Questions?
Catalog NumberColor
FKS11911583Clear
Catalog number FKS11911583
Price (SGD)
319.00
PK
Add to cart
Color:
Clear
Price (SGD)
319.00
PK
Add to cart
Store your samples with confidence in convenient Thermo Scientific™ National™ Sample Storage Assembled Screw Vial Kits.
  • Superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass vials
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
  • Pack of 100
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorWhite
DescriptionScrew Vial Assembled Kit, Solid-top Cap
ColorClear
MaterialGlass
Volume (Metric)20 mL
No. per Pack100
Closure MaterialPolypropylene
Closure TypeScrew Cap
Closure Size24-400
Unit SizePK