Manufactured from Pyrex™ borosilicate glass with PTFE key, for superior chemical resistance. Pyrex™ Borosilicate Glass Stopcock with Key are made to a taper of 1:10 giving a combination of performance and ease of operation.
Have Questions?
Catalog Number
Diameter (Metric) Side Arm
Plug Bore
Type
FKS11910955
10 mm
2.5 mm
Single Bore Stopcock
Catalog number FKS11910955
Price (SGD)
762.00
PK
Add to cart
Diameter (Metric) Side Arm:
10 mm
Plug Bore:
2.5 mm
Type:
Single Bore Stopcock
Price (SGD)
762.00
PK
Add to cart
Manufactured from Pyrex™ borosilicate glass with PTFE key, for superior chemical resistance. Pyrex™ Borosilicate Glass Stopcock with Key are made to a taper of 1:10 giving a combination of performance and ease of operation.
Barrels: highly polished
Keys: self-lubricating, avoiding the need for stopcock grease
Keys: fully interchangeable and include plastic screw thread retaining device