Tychem 6000 F Sleeves
Tychem 6000 F  Sleeves
Actual product may vary
Tychem 6000 F  Sleeves
Tychem 6000 F  Sleeves
Fisherbrand
Tychem 6000 F Sleeves
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tychem™ 6000 F Sleeves come with stitched and over-taped seams for protection and strength.
Have Questions?
Catalog number FKS11905864
Price (SGD)
-
  • Antistatic treatment
  • Wide elastics at cuff and upper arm
  • Excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Specifications
ConstructionHeavy-duty Tyvek substrate
Product TypeChemical-resistant Sleeve
SizeUniversal
MaterialTyvek Substrate
ColorGray
Seam TypeStitched, Over-taped
Cuff StyleElastic
FinishPolymeric barrier film
Certifications/Compliance(EU) 2016/425, Category III, Type PB [3], EN 14126, EN 1149-5
Length (Metric)500 mm
Weight (Metric)5 kg
Unit SizePK