For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
Have Questions?
Change view
Catalog Number
Length (Metric)
FKS11899601
590 mm
FKS11869701
740 mm
2 Options
Catalog number FKS11899601
Price (SGD)
615.00
Each
Length (Metric):
590 mm
Price (SGD)
615.00
Each
Manufactured from stainless steel
For handling a wide variety of crucibles, especially in muffle furnaces
Operated by thumb action with one hand
Specifications
For Use With (Application)For handling a wide variety of crucibles and to protect the hand against radiation from the furnace