Sterileware™ V-shaped Sampling Spatula, Sterile
Sterileware™ V-shaped Sampling Spatula, Sterile
Actual product may vary
Fisher Catalogue
Sterileware™ V-shaped Sampling Spatula, Sterile
Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
Have Questions?
Catalog NumberLength (Metric)
FKS1189782222.9 cm
Catalog number FKS11897822
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
22.9 cm
Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
  • Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze.
  • Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18 mm).
  • Sterileware (16-241-210, 14-385-20) available in 9 and 14 in. models.
  • High-impact polystyrene.
  • Individually packaged.
  • Sterilized by gamma irradiation.
  • Lot stamped, shipped with Certificate of Processing.
  • Nonsterile Model (03-421-149) 9 in. spatula.
Specifications
SterilitySterile
AutoclavableNot Autoclavable
ShapePointed End
DisposableSingle-use
Product TypeSterileware™ Sampling Spatula
For Use With (Equipment)Containers, bags and drums
For Use With (Application)Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections and for piercing Plastic-lined Paper bags or similar package
Length (English)9 in.
Width (English)0.75 in.
Width (Metric)1.9 cm
Dimensions (L x W)9 x 0.75 in. (22.9 x 1.9 cm)
Includes6 in. L V-shaped channel
Length (English) Handle2.99 in.
Length (Metric) Handle7.6 cm
MaterialPolystyrene
Length (Metric)22.9 cm
Unit SizePK