Wire Glove Box Holder
Wire Glove Box Holder
Actual product may vary
Fisherbrand
Wire Glove Box Holder
Hold three boxes of gloves in a wall-mounted holder. Fisherbrand™ Wire Glove Box Holder helps free up benchtop space and is compatible with most major glove brands. Constructed of sturdy, epoxy-coated steel wire for an economical, easy-to-clean solution.
Have Questions?
Catalog NumberDepth (Metric)
FKS11895843108 mm
Catalog number FKS11895843
Price (SGD)
149.00
EA
Add to cart
Depth (Metric):
108 mm
Price (SGD)
149.00
EA
Add to cart
  • Keep gloves organized and ready for immediate use
  • Free up benchtop space by mounting holders on wall for easy dispensing
  • Vertical three-box holder fits most major brands of glove boxes
  • Wall mounting screws included
Specifications
Product TypeGlove Box Holder
ColorWhite
Depth (Metric)108 mm
Dimensions (D x W x H) Interior108 x 250 x 454 mm
Width (Metric)250 mm
Height (Metric)454 mm
HoldsGloves
MaterialEpoxy-coated Steel
Capacity3 Boxes
IncludesWall mounting screws
Quantity1 Ea.
Unit SizeEA