Bandido Safety Glasses
Bandido Safety Glasses
Actual product may vary
Fisherbrand
Bandido Safety Glasses
Genuine protection and high value in an inexpensive model.
Have Questions?
Catalog NumberLens TintLensType
FKS11895834Clear
Catalog number FKS11895834
Price (SGD)
22.00
EA
Add to cart
Lens Tint:
Clear
Price (SGD)
22.00
EA
Add to cart
  • An inexpensive model for genuine protection.
  • Made entirely of highly-resistant polycarbonate, BANDIDO is the perfect value option.
  • Wrap-around fit.
  • EN 166: Guaranteed minimum resistance of the protective eyewear against everyday risks including dropping the protective eyewear onto the ground, ageing by light exposure, exposure to heat or corrosion etc.
  • F, Mechanical risk - Low energy impact: Low energy impact, resists a 6 mm, 0.86 g ball at 45 m/s.
  • T, Mechanical risk- High speed particles at extreme temperatures: The letter T, immediately after the mechanical strength symbol, authorizes use for high-speed particles at extreme temperatures.
  • Optical class: Class 1, continuous work, worn permanently.
  • CE, European standard: Frame marked to indicate meeting CE standard.
Specifications
Lens CoatingAnti-fog, Scratch-resistant
DescriptionModel BANCI
Certifications/Compliance (Europe)EN166, EN170, Mechanical risk F/T, Optical class 1, CE
Lens TintClear
Product TypeSafety Spectacles
For Use With (Application)Protection, Continuous Wear
IncludesNeck cord
No. per Pack1 Ea.
Weight (Metric)28 g
MaterialPolycarbonate
Lens MaterialPolycarbonate
Frame MaterialPlastic
UV ProtectionYes
Frame TypeWrap-around
Unit SizeEA