Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) |
---|---|
FKS11895762 | 0.5 to 5 mL |
FISFBE00002 | 0.2 to 2 μL |
FKS11815762 | 0.2 to 2 μL |
FKS11825762 | 0.5 to 5 μL |
FISFBE00010 | 1 to 10 μL |
FKS11835762 | 1 to 10 μL |
FISFBE00020 | 2 to 20 μL |
FKS11845762 | 2 to 20 μL |
FISFBE00050 | 5 to 50 μL |
FKS11855762 | 5 to 50 μL |
FISFBE00100 | 10 to 100 μL |
FKS11865762 | 10 to 100 μL |
FISFBE00200 | 20 to 200 μL |
FKS11875762 | 20 to 200 μL |
FISFBE10000 | 1 to 10 mL |
FKS11805772 | 1 to 10 mL |
FISFBE05000 | 0.5 to 5 mL |
FISFBE01000 | 100 to 1000 μL |
FKS11885762 | 100 to 1000 μL |