1x1 image pixel for data collection
Metalware Range™ Stainless Steel Petri Dish Sterilizing Box
Metalware Range™ Stainless Steel Petri Dish Sterilizing Box
Actual product may vary
Fisherbrand
Metalware Range™ Stainless Steel Petri Dish Sterilizing Box
Nickel Electro™ Metalware Range™ Stainless Steel Petri Dish Sterilizing Box holds 10 petri dishes in a convenient removable stainless carrier.
Have Questions?
Catalog NumberLength (Metric)
FKS11894483254 mm
Catalog number FKS11894483
Price (SGD)
485.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Length (Metric):
254 mm
Price (SGD)
485.00
EA
Add to cart
  • Manufactured from stainless steel
  • Non-magnetic
  • Bright finish
  • Can be steam sterilised at 120°C and withstand dry heat temperatures of up to 180°C
Specifications
AutoclavableAutoclavable
Dimensions (L x W)254 x 115 mm
MaterialStainless Steel
SterilityNon-sterile
For Use With (Application)For Transportation and Sterilization
QuantityEach
Weight (Metric)0.76 kg
Length (Metric)254 mm
TypeSterilizing Box
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link