Whatman™ Acid/Alkali Test Papers
Whatman™ Acid/Alkali Test Papers
Actual product may vary
Fisherbrand
Whatman™ Acid/Alkali Test Papers
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor IndicatorFormat
FKS11894463Litmus BlueStrip
FKS11804473Litmus RedStrip
Catalog number FKS11894463
Price (SGD)
489.00
PK
Add to cart
Color Indicator:
Litmus Blue
Format:
Strip
Price (SGD)
489.00
PK
Add to cart
Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.
  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3
Specifications
Color IndicatorLitmus Blue
pH Range5 to 8
FormatStrip
Quantity10 x 10 Books of 20 Strips
TypepH Test Paper
Length (Metric)60 mm
Width (Metric)10 mm
Unit SizePK