Stainless Steel Hinged Gable Lid
Stainless Steel Hinged Gable Lid
Actual product may vary
Fisherbrand
Stainless Steel Hinged Gable Lid
Nickel Electro™ Clifton™ Stainless Steel Hinged Gable Lid minimises heat loss and reduces evaporation while maintaining bath temperature stability.
Have Questions?
Catalog NumberFor Use With (Equipment)
FKS11894313For 22 and 28 L Baths
Catalog number FKS11894313
Price (SGD)
1,575.00
EA
Add to cart
For Use With (Equipment):
For 22 and 28 L Baths
Price (SGD)
1,575.00
EA
Add to cart
  • Designed allowing full use of whole width of a water bath with the lowest height of the gable being 45mm above the bath.
  • Test tube racks or flasks can be placed across the whole width of the bath without fouling the lid maximising the bath working area.
  • Designed with customer in mind ensuring where condensation accumulates in use there are no drip points which could spoil work in open flasks in your water bath.
  • Lid fits into the tank ensuring all condensation runs back into the bath and not down the side onto your bench top.
  • Reduces energy consumption and running costs for your water bath.
  • Insulated handle.
  • The hinged lid has the added advantage where bench space is limited, the lid hinges back to the vertical position to access the bath where as a conventional lid requires lifting off the bath and placing on your bench top.
  • With continuous sampling work a conventional lid has to be removed frequently to access samples/work this hinged lid is designed with this in mind.
  • The Lid hinges back to just over vertical position as in image above.
Specifications
DescriptionHinged stainless steel gable lid
For Use With (Equipment)For 22 and 28 L Baths
Warranty3 Years
Unit SizeEA