Check Order Status
Translucent Polypropylene T-shape Tubing Connector
Actual product may vary
Fisherbrand
Translucent Polypropylene T-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with T-style side arm.
Have Questions?
Change view
Catalog Number
Tubing Size
FKS11893920
4 to 5 mm tubing I.D.
FKS11813930
8 to 9 mm tubing I.D.
FKS11803930
6 to 7 mm tubing I.D.
3 Options
Catalog number
FKS11893920
Price (SGD)
80.00
PK
Add to cart
Tubing Size:
4 to 5 mm tubing I.D.
Price (SGD)
80.00
PK
Add to cart
Translucent polypropylene construction
Accommodates 3 tubes of the same diameter
Specifications
Material
Polypropylene
Overall Length (Metric)
30 mm
For Use With
4 to 5 mm tubing I.D.
Packaging Quantity
20 Pack
Tubing Size
4 to 5 mm tubing I.D.
Shape
T-shape
Includes
23 mm side arm
Color
Translucent
Product Type
Tubing Connector