Class B Polypropylene Short Form Measuring Cylinders
Class B Polypropylene Short Form Measuring Cylinders
Actual product may vary
Fisherbrand
Class B Polypropylene Short Form Measuring Cylinders
Use these cylinders for food industry and volume measurement applications. VITLAB™ Class B Polypropylene Short Form Measuring Cylinders feature easy to read, raised scale and ring marks at the primary scale points.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS11893662500 mL10 mL
Catalog number FKS11893662
Price (SGD)
216.00
PK
Add to cart
Capacity (Metric):
500 mL
Graduations:
10 mL
Price (SGD)
216.00
PK
Add to cart
  • Highly transparent polypropylene graduated cylinder
  • Calibrated (In)
  • Class B tolerances according to DIN 12681 / ISO 6706.
  • Thermal stress up to 80°C does not cause tolerance limits to be permanently exceeded
Specifications
Capacity (Metric)500 mL
MaterialPolypropylene
Tolerance±10 mL
ClassB
CalibrationTo Contain
ScaleRaised Scale and Ring Marks at the Primary Scale Points
Diameter (Metric)67 mm
Graduations10 mL
Limit of Error±10mL
ShapeRound
Height (Metric)218 mm
TypeGraduated Cylinder
Unit SizePK