Acrylic Safety Glasses Dispenser
Acrylic Safety Glasses Dispenser
Actual product may vary
Fisher Catalogue
Acrylic Safety Glasses Dispenser
Access safety accessories with ease. Fisherbrand™ Acrylic Safety Glasses Dispenser holds protective goggles, other safety items and personal items to keep them safe and clean. They are ergonomically angled for easy retrieval. Transparent blue dispensers are available in two sizes.
Have Questions?
Catalog NumberSize
FKS11893460Large
Catalog number FKS11893460
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Large
  • Translucent acrylic dispensers provide easy access to safety accessories
  • Ergonomically angled for easy dispensing
  • Hinged lid protects glasses from dust and debris
  • Flip open top for filling with tilt-open bottom for dispensing
  • Set on counter or wall mount (screws included) for convenient access
  • Large dispenser holds approximately 20 pairs of glasses
    Small dispenser holds approximately 10 pairs of glasses
Specifications
Width (Metric)203 mm
Depth (Metric)406 mm
ColorBlue
Product TypeDispenser
For Use With (Equipment)Safety Glasses
IncludesWall mounting screws
Length (Metric)203 mm
No. per Pack1 Ea.
SizeLarge
MaterialAcrylic
Unit SizeEA