Octagonal Magnetic Stir Bar Assortment with Retainer
Octagonal Magnetic Stir Bar Assortment with Retainer
Actual product may vary
Fisher Catalogue
Octagonal Magnetic Stir Bar Assortment with Retainer
Provides greater surface area and added turbulence when compared to smooth cylindrical bars. Fisherbrand™ Octagonal Magnetic Stir Bar Assortments with Restrainers have an integral pivot ring on each bar to aid in reducing friction and chattering.
Have Questions?
Catalog number FKS11893210
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Alnico magnets encapsulated in FDA grade low friction coating
  • Restrainer fits comfortably in one hand
  • Eliminates splashing hazards when pouring
Specifications
Certifications/ComplianceISO 9001:2008
Dimensions (L x W x H)12.7 x 3.175, 12.7 x 7.92, 38.1 x 7.92, 50.8 x 7.92mm
IncludesIntegral Pivot Rings, Spinbar Restrainer
CoatingPTFE
MaterialPTFE
Quantity1/Ea.
ShapeOctagon
TypeMagnetic Stir Bar, Micro
Unit SizeEA