▸
Pipet Products
▸
Pipet Bulbs
▸
Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Actual product may vary
Fisherbrand
Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Have Questions?
Catalog Number
Color
FKS11890950
Red
Catalog number
FKS11890950
Price (SGD)
21.00
Each
Add to cart
Color:
Red
Price (SGD)
21.00
Each
Add to cart
Available in a range of colors
With ca. 60mL volume suitable for pipettes from 2 to 50mL
Pipette media must not enter pipette filler; it may cause valve malfunction
Specifications
Color
Red
Material
Rubber
Type
Pipette Filler
Volume (Metric)
60 mL
For Use With (Equipment)
Can be used on bulb or cylindrical pipettes from 2mL to 50mL
Includes
Three Glass ball valves
Quantity
1 Each