Check Order Status
Cylindrical PTFE or Glass Stir Bars
Actual product may vary
Fisher Catalogue
Cylindrical PTFE or Glass Stir Bars
Alnico V magnet core encapsulated by PTFE or glass . Fisherbrand™ Cylindrical Stir Bars are autoclavable.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11888932
15 mm
6 mm
FKS11868942
40 mm
8 mm
FKS11858922
20 mm
6 mm
FKS11868922
25 mm
6 mm
4 Options
Catalog number
FKS11888932
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
15 mm
Diameter (Metric):
6 mm
Stable, general-purpose magnetic stir bar
Smooth, round profile for a wide variety of applications
Autoclavable
Specifications
Autoclavable
Autoclavable
Certifications/Compliance
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color
White
Description
PTFE encapsulated Alnico V magnet core
For Use With (Equipment)
Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)
6 mm
Length (Metric)
15 mm
Quantity
10/Pk.
Shape
Cylinder