Silberbrand™ Borosilicate Glass Class B Volumetric Flask
Silberbrand™ Borosilicate Glass Class B Volumetric Flask
Actual product may vary
Fisher Catalogue
Silberbrand™ Borosilicate Glass Class B Volumetric Flask
Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Silberbrand™ Borosilicate Glass Class B Volumetric Flask includes polypropylene stopper. White enamel DE-M marking provides quick volume check.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeLimit of Error
FKS11888351250 mL14/23±0.25 mL
FKS1185835150 mL12/21±0.09 mL
Catalog number FKS11888351
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Closure Size:
14/23
Limit of Error:
±0.25 mL
  • Manufactured from Borosilicate Glass
  • Complies with DIN EN ISO 1042
  • Class B
  • Calibrated to contain (TC, In.)
  • Includes polypropylene stopper
  • White enamel DE-M marking provides quick volume check
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolypropylene
ColorClear
Capacity (Metric)250 mL
MaterialBorosilicate Glass
Limit of Error±0.25 mL
No. per Case2 Piece
Tolerance±0.25 mL
ClassB
Closure Size14/23
Unit SizePK