Search Thermo Fisher Scientific
Catalog Number | Includes |
---|---|
FKS11888203 | Plastic Screw Head |
FKS11517722 | Metal Screw Head |
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
Note:
The clamp and boss head are sold separately.