Polypropylene Cryoboxes
Polypropylene Cryoboxes
Actual product may vary
Fisher Catalogue
Polypropylene Cryoboxes
Use these polypropylene boxes with grid inserts for ultra-low temperature storage. Fisherbrand™ Polypropylene Cryoboxes are ideal for sample storage and transportation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHolds
FKS11886903Red3 to 5 mL
FKS11856903Blue1 to 2 mL
FKS11806913Blue3 to 5 mL
FKS11846903Green1 to 2 mL
FKS11856893Natural1 to 2 mL
FKS11836903Red1 to 2 mL
Catalog number FKS11886903
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Red
Holds:
3 to 5 mL
  • Autoclavable, temperature resistant from -90°C to +121°C
  • High quality polypropylene
  • Available in natural and various colors
  • 81 places, 9 x 9 grid, numerically coded
  • Robust hinge with safe snap-on lid
  • Stackable for safe transportation
  • Drain bores at the bottom for dew liquids
  • Removable grid inserts
  • Can also be used for larger vessels by removing the grid inserts For sample storage and transportation.
Specifications
Capacity (Metric)5 mL
Closure TypeSnap-On Lid
ColorRed
MaterialPolypropylene
ShapeSquare
TypeCryo Box
Holds3 to 5 mL
IncludesStackable Digital Identification, Translucent Lid Hinge
Max. Temperature (Metric)121°C
Min. Temperature (Metric)-90°C
Quantity5 Pack
Length (Metric)133 mm
Height (Metric)75 mm
Weight (Metric)0.8 kg
Unit SizePK