Pressureless LDPE Spray Bottles with Long-life Hand Pump
Pressureless LDPE Spray Bottles with Long-life Hand Pump
Actual product may vary
Fisherbrand
Pressureless LDPE Spray Bottles with Long-life Hand Pump
Manufactured from polypropylene, polyethylene and stainless steel. Buerkle™ Pressureless LDPE Spray Bottles spray 1.2ml ±0.1 volume per stroke. Suitable for agricultural pesticides and detergents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Neck ID (Metric)
FKS118868401000 mL17.5 mm
FKS10190071500 mL18 mm
Catalog number FKS11886840
Price (SGD)
33.00
PK
Add to cart
Capacity (Metric):
1000 mL
Neck ID (Metric):
17.5 mm
Price (SGD)
33.00
PK
Add to cart
  • Media-contacting parts PP, PE, and stainless steel
  • Bottle made of LDPE
  • Nozzle diameter 0.6mm
  • Spray volume per stroke: 1.2ml ±0.1
  • Adjustable spray jet
  • Temperature max. 30°C
  • The functioning of individual sprayers is tested during manufacture
Specifications
Capacity (Metric)1000 mL
For Use With (Application)Industry, Laboratories, Production, Leisure, Household and Suitable for Agricultural Pesticides and Detergents
IncludesPatented venting valve, long-life hand pump
MaterialLDPE
Neck ID (Metric)17.5 mm
Temperature Range (Metric)30°C
ShapeRound
Height (Metric)295 mm
Closure TypeSpray
Unit SizePK