Clifton™ Stainless Steel Unstirred Digital Water Baths
Clifton™ Stainless Steel Unstirred Digital Water Baths
Actual product may vary
Clifton™ Stainless Steel Unstirred Digital Water Baths
Clifton™ Stainless Steel Unstirred Digital Water Baths
Fisher Catalogue
Clifton™ Stainless Steel Unstirred Digital Water Baths
Get optimum water bath results with this stainless steel unstirred digital water bath featuring a digital PID temperature control for precision. Ideal for meeting quality control requirements, this Clifton™ Stainless Steel Unstirred Digital Water Bath features digital PID temperature control for precision.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1188536056 L
FKS1141950914 L
FKS1143950928 L
FKS114095098 L
Catalog number FKS11885360
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
56 L
  • Designed for immersion of flasks/bottles/racks in a highly stable temperature environment
  • Designed for operating with low liquid levels suitable for small bottles, tubes and microplates
  • Corrosion resistant stainless steel working environment
  • Durable powder coated exterior, chemical resistant and now
  • Features anti-bacterial paint finish that inhibits the growth of bacteria
  • For heating samples in microcentrifuge tubes and molecular biology applications
  • Low height design maximizing the working height in the bath
Specifications
Controller TypePID Digital, LED Display
DescriptionDigital Water Bath
Capacity (Metric)56 L
Certifications/ComplianceCE Approved
Temperature Range5°C to 99°C
For Use With (Application)Providing a Stable Temperature Environment Ideal for Meeting Quality Control Requirements Featuring
MaterialStainless Steel
ModelNE2-56 D
Temperature Resolution±0.1°C
Temperature Uniformity±0.1°C
Warranty3 Years
Dimensions (D x W x H)540 x 361 x 290 mm
Dimensions (D x W x H) Interior500 x 325 x 200 mm
Width (Metric)361 mm
Width (Metric) Interior325 mm
Height (Metric)290 mm
Height (Metric) Interior200 mm
Depth (Metric)540 mm
Depth (Metric) Interior500 mm
Weight (Metric)12.5 kg
Voltage230 V
Wattage2500 W